Läs mer
4,5,6,7-Tetrabromo-1H-benzotriazole, TRC
Produktkod. 30410994
Change view
Kvantitet:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30410994 5 mg
30410450 50 mg
30420920 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30410994 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2913255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17374-26-4
Molekylformel C6 H Br4 N3
Molekylvikt (g/mol) 434.71
Synonym TBB
IUPAC-namn 4,5,6,7-tetrabromo-1H-benzotriazole
LEDER Brc1c(Br)c(Br)c2[nH]nnc2c1Br
Specifikationer
Kemiskt namn eller material 4,5,6,7-Tetrabromo-1H-benzotriazole
Kvantitet 5 mg
InChI formel InChI=1S/C6HBr4N3/c7-1-2(8)4(10)6-5(3(1)9)11-13-12-6/h(H,11,12,13)
Formel vikt 430.6904