Läs mer
4,5,6,7-Tetrabromobenzimidazole, TRC
Produktkod. 30415099
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30415099 100 mg
30410734 250 mg
30412615 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30415099 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT291150100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 577779-57-8
Molekylformel C7 H2 Br4 N2
Molekylvikt (g/mol) 433.72
IUPAC-namn 4,5,6,7-tetrabromo-1H-benzimidazole
LEDER Brc1c(Br)c(Br)c2[nH]cnc2c1Br
Specifikationer
Kemiskt namn eller material 4,5,6,7-Tetrabromobenzimidazole
Kvantitet 100 mg
InChI formel InChI=1S/C7H2Br4N2/c8-2-3(9)5(11)7-6(4(2)10)12-1-13-7/h1H,(H,12,13)
Formel vikt 429.6951