Learn More
4,5,6,7-Tetrabromophenolphthalein (>80%), TRC
Specifications
Specifications
| Chemical Name or Material | 4,5,6,7-Tetrabromophenolphthalein |
| CAS | 13027-28-6 |
| Molecular Formula | C20 H10 Br4 O4 |
| Quantity | 100 mg |
| Synonym | 4,5,6,7-tetrabromo-3,3-bis(4-hydroxyphenyl)-1(3H)-Isobenzofuranone, 4,5,6,7-tetrabromo-Phenolphthalein (6CI,7CI,8CI), 4,5,6,7-Tetrabromo-3,3-bis(4-hydroxyphenyl)-1(3H)-isobenzofuranone, 4,5,6,7-Tetrabromo-3,3-bis(4-hydroxyphenyl)phthalide, Phenoltetrabromphthalein |
| InChI Formula | InChI=1S/C20H10Br4O4/c21-15-13-14(16(22)18(24)17(15)23)20(28-19(13)27,9-1-5-11(25)6-2-9)10-3-7-12(26)8-4-10/h1-8,25-26H |
| SMILES | Oc1ccc(cc1)C2(OC(=O)c3c(Br)c(Br)c(Br)c(Br)c23)c4ccc(O)cc4 |
| IUPAC Name | 4,5,6,7-tetrabromo-3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one |
| Molecular Weight (g/mol) | 633.907 |
| Formula Weight | 629.731 |