Läs mer
4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1-formylpiperidine, Mikromol™
Produktkod. 17960886
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17960886 100 mg
17901026 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17960886 Leverantör Mikromol Leverantörsnummer MM0107.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1-formylpiperidine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1-formylpiperidine
CAS 1346598-11-5
Analyt- eller komponentnamn 4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1-formylpiperidine
Frakt skick Room Temperature
Molekylformel C13 H14 Cl N3 O2
Certifikat och efterlevnad ISO 9001
Synonym Domperidone Imp. B (EP), 4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1-formylpiperidine, Domperidone Impurity B
InChI formel InChI=1S/C22H28N2O2.ClH/c1-2-22(25)23(20-11-7-4-8-12-20)21-14-17-24(26,18-15-21)16-13-19-9-5-3-6-10-19;/h3-12,21H,2,13-18H2,1H3;1H
LEDER Clc1ccc2N(C3CCN(CC3)C=O)C(=O)Nc2c1
IUPAC-namn 4-(5-chloro-2-oxo-3H-benzimidazol-1-yl)piperidine-1-carbaldehyde
Molekylvikt (g/mol) 279.72
Kvantitet 100 mg
Formel vikt 279.0775 g/mol