Läs mer
4,5-Diamino-1-(2-hydroxyethyl)pyrazole Sulfate, TRC
Produktkod. 30358611
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358611 100 mg
30359745 500 mg
30350746 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30358611 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD415145100MG
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 155601-30-2
Molekylformel C5H10N4O . H2SO4
Molekylvikt (g/mol) 240.23
Synonym 1-(2-Hydroxyethyl)-4,5-diaminopyrazole Sulfate, 4,5-Diamino-1-(2-hydroxyethyl)-1H-pyrazole Sulfate, Jarocol AHP, Rodol HDAP, 4,5-Diamino-1H-pyrazole-1-ethanol Sulfate (1:1)
IUPAC-namn 2-(4,5-diamino-1H-pyrazol-1-yl)ethan-1-ol sulfate
LEDER Nc1c(N)n(CCO)nc1.OS(=O)(O)=O
Specifikationer
Kemiskt namn eller material 4,5-Diamino-1-(2-hydroxyethyl)pyrazole Sulfate
Kvantitet 100 mg
InChI formel InChI=1S/C5H10N4O.H2O4S/c6-4-3-8-9(1-2-10)5(4)7;1-5(2,3)4/h3,10H,1-2,6-7H2;(H2,1,2,3,4)
Formel vikt 240.0528