Läs mer
4,5-Diamino-6-methylpyrimidine Hydrochloride, TRC
Produktkod. 30359339
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30359339 100 mg
1 options
Produktkod. 30359339 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD416376100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4,5-Diamino-6-methylpyrimidine Hydrochloride
CAS 1797132-20-7
Molekylformel C5H8N4 • HCl
Kvantitet 100 mg
Synonym 4,5-Diamino-6-methyl-pyrimidine Hydrochloride, NSC 122008 Hydrochloride
InChI formel InChI=1S/C5H8N4.ClH/c1-3-4(6)5(7)9-2-8-3;/h2H,6H2,1H3,(H2,7,8,9);1H
LEDER CC1=C(C(=NC=N1)N)N.Cl
Molekylvikt (g/mol) 124.143646
Formel vikt 160.0516