missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4,5-Dichloro-2-nitrobenzotrifluoride, 98%

Brand:  Alfa Aesar™ H61893.14

Additional Details : CAS Number : 50594-31-5 Weight : 0.02500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15498478

  • 1708.51 SEK / 25g


Estimated Shipment date
from Supplier 15-07-2019
Add to basket

Specifications

Chemical Identifiers

50594-31-5
259.993
MMUARSWOJRDXBV-UHFFFAOYSA-N
2736844
C1=C(C(=CC(=C1Cl)Cl)[N+](=O)[O-])C(F)(F)F
C7H2Cl2F3NO2
MFCD00236666
1,2-dichloro-4-nitro-5-trifluoromethyl benzene, 4,5-dichloro-2-nitrobenzotrifluoride, 3,4-dichloro-6-nitrobenzotrifluoride, benzene, 1,2-dichloro-4-nitro-5-trifluoromethyl, 4,5-dichloro-2-trifluoromethyl nitrobenzene, pubchem4863, ksc915a8h, 4,5-dichloro-2-nitrobenzenetrifluoride, 2-nitro-4,5-dichlorobenzotrifluoride, 4,5-dichloro-1-nitro-2-trifluoromethyl benzene
1,2-dichloro-4-nitro-5-(trifluoromethyl)benzene

Specifications

4,5-Dichloro-2-nitrobenzotrifluoride
Yellow
25g
C7H2Cl2F3NO2
UN2811
MMUARSWOJRDXBV-UHFFFAOYSA-N
1,2-dichloro-4-nitro-5-(trifluoromethyl)benzene
2736844
Crystalline Solid
1.600g/mL
36°C to 38°C
50594-31-5
MFCD00236666
1,2-dichloro-4-nitro-5-trifluoromethyl benzene, 4,5-dichloro-2-nitrobenzotrifluoride, 3,4-dichloro-6-nitrobenzotrifluoride, benzene, 1,2-dichloro-4-nitro-5-trifluoromethyl, 4,5-dichloro-2-trifluoromethyl nitrobenzene, pubchem4863, ksc915a8h, 4,5-dichloro-2-nitrobenzenetrifluoride, 2-nitro-4,5-dichlorobenzotrifluoride, 4,5-dichloro-1-nitro-2-trifluoromethyl benzene
C1=C(C(=CC(=C1Cl)Cl)[N+](=O)[O-])C(F)(F)F
259.993
260
98%
Videos
SDS