Läs mer
4,5-Dimethyl-3-hydroxy-2(5H)-furanone-13C2, TRC
Produktkod. 17943195
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17943195 1 mg
17943015 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17943195 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4736031MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 156420-66-5
Molekylformel 13C2 C4 H8 O3
Molekylvikt (g/mol) 130.11
Synonym 2(5H)-Furanone-5-13C, 3-hydroxy-4-methyl-5-(methyl-13C)- (9CI), 4,5-Dimethyl-3-hydroxy-2(5H)-furanone-13C2, 3-Hydroxy-4-methyl-5-methyl-13C-2(5H)-furanone-5-13C, 4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one-13C2, 4,5-Dimethyl-3-hydroxy-2(5H)-furanone-13C2 (5,5-methyl-13C2)
IUPAC-namn 4-hydroxy-3-methyl-2-(1^{13}C)methyl-(2^{13}C)2H-furan-5-one
LEDER [13CH3][13CH]1OC(=O)C(=C1C)O
Specifikationer
Kemiskt namn eller material 4,5-Dimethyl-3-hydroxy-2(5H)-furanone 5,5-methyl-13C2
Kvantitet 1 mg
InChI formel InChI=1S/C6H8O3/c1-3-4(2)9-6(8)5(3)7/h4,7H,1-2H3/i2+1,4+1
Formel vikt 130.0541