Läs mer
4,5Alpha-Dihydronorethisterone, TRC
Produktkod. 30137899
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137899 10 mg
30009659 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137899 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44951010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 52-79-9
Molekylformel C20 H28 O2
Molekylvikt (g/mol) 300.44
Synonym 19-Norpregn-20-yn-3-one, 17-hydroxy-, (5α,17α)-, 19-Nor-5α,17α-pregn-20-yn-3-one, 17-hydroxy- (6CI,7CI,8CI), (5α,17α)-17-Hydroxy-19-norpregn-20-yn-3-one, 17α-Ethynyl-17β-hydroxy-5α-estran-3-one, 17α-Ethynyl-5α-estran-17β-ol-3-one, 17β-Hydroxy-17α-ethynyl-5α-estran-3-one, 17β-Hydroxy-19-nor-5α,17α-pregn-20-yn-3-one, 5α-Dihydronorethindrone, 5α-Dihydronorethisterone, NSC 85401, STS 737, 4,5α-Dihydronorethisterone
IUPAC-namn (5S,8R,9R,10S,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,4,5,6,7,8,9,10,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-3-one
LEDER C[C@]12CC[C@@H]3[C@H]4CCC(=O)C[C@@H]4CC[C@H]3[C@@H]1CC[C@@]2(O)C#C
Specifikationer
Kemiskt namn eller material 4,5alpha-Dihydronorethisterone
Kvantitet 10 mg
InChI formel InChI=1S/C20H28O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,13,15-18,22H,4-12H2,2H3/t13-,15-,16+,17+,18-,19-,20-/m0/s1
Formel vikt 300.2089