Läs mer
4',6,7-Trihydroxyisoflavone, TRC
Produktkod. 30009436
Change view
Kvantitet:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009436 100 mg
30006102 50 mg
30012700 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009436 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT795360100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17817-31-1
Molekylformel C15 H10 O5
Molekylvikt (g/mol) 270.24
Synonym Demethyltexasin, 6-Hydroxydaidzein, Desmethylglycitein, Soybean factor 2
IUPAC-namn 6,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one
LEDER Oc1ccc(cc1)C2=COc3cc(O)c(O)cc3C2=O
Specifikationer
Kemiskt namn eller material 4',6,7-Trihydroxyisoflavone
Kvantitet 100 mg
InChI formel InChI=1S/C15H10O5/c16-9-3-1-8(2-4-9)11-7-20-14-6-13(18)12(17)5-10(14)15(11)19/h1-7,16-18H
Formel vikt 270.0528