Läs mer
4-[6-[(8-Cyclopentyl-6-ethenyl-7,8-dihydro-5-methyl-7-oxopyrido[2,3-d]pyrimidin-2-yl)amino]-3-pyridinyl]-1-Piperazinecarboxylic Acid 1,1-Dimethylethyl Ester, TRC
Produktkod. 30357916
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30357916 1 mg
30364512 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30357916 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9925631MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1941177-45-2
Molekylformel C29H37N7O3
Molekylvikt (g/mol) 531.66
Synonym 1-Piperazinecarboxylic acid, 4-[6-[(8-cyclopentyl-6-ethenyl-7,8-dihydro-5-methyl-7-oxopyrido[2,3-d]pyrimidin-2-yl)amino]-3-pyridinyl]-, 1,1-dimethylethyl ester (ACI), 1,1-Dimethylethyl 4-[6-[(8-cyclopentyl-6-ethenyl-7,8-dihydro-5-methyl-7-oxopyrido[2,3-d]pyrimidin-2-yl)amino]-3-pyridinyl]-1-piperazinecarboxylate (ACI)
IUPAC-namn tert-butyl 4-[6-[(8-cyclopentyl-6-ethenyl-5-methyl-7-oxopyrido[2,3-d]pyrimidin-2-yl)amino]pyridin-3-yl]piperazine-1-carboxylate
LEDER CC1=C(C=C)C(=O)N(C2CCCC2)c3nc(Nc4ccc(cn4)N5CCN(CC5)C(=O)OC(C)(C)C)ncc13
Specifikationer
Kemiskt namn eller material 4-[6-[(8-Cyclopentyl-6-ethenyl-7,8-dihydro-5-methyl-7-oxopyrido[2,3-d]pyrimidin-2-yl)amino]-3-pyridinyl]-1-Piperazinecarboxylic Acid 1,1-Dimethylethyl Ester
Kvantitet 1 mg
InChI formel InChI=1S/C29H37N7O3/c1-6-22-19(2)23-18-31-27(33-25(23)36(26(22)37)20-9-7-8-10-20)32-24-12-11-21(17-30-24)34-13-15-35(16-14-34)28(38)39-29(3,4)5/h6,11-12,17-18,20H,1,7-10,13-16H2,2-5H3,(H,30,31,32,33)