Learn More
4-(6-Amino-3-pyridyl)-1-Boc-piperazine, 97%, Thermo Scientific™
1005.00 SEK - 10980.24 SEK
Chemical Identifiers
CAS | 571188-59-5 |
---|---|
Molecular Formula | C14H22N4O2 |
Molecular Weight (g/mol) | 278.36 |
MDL Number | MFCD11594962 |
InChI Key | RMULRXHUNOVPEI-UHFFFAOYSA-N |
Synonym | tert-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, 1-boc-4-6-aminopyridin-3-yl piperazine, tert-butyl 4-6-amino-3-pyridyl piperazine-1-carboxylate, 1-boc-4-6-amino-3-pyridyl piperazine, 4-6-amino-3-pyridyl-1-boc-piperazine, 1-piperazinecarboxylic acid, 4-6-amino-3-pyridinyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-6-amino-3-pyridyl piperazine, t-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, tert-butyl4-6-aminopyridin-3-yl piperazine-1-carboxylate, 4-6-amino-3-pyridyl piperazine-1-carboxylic acid tert-butyl ester |
PubChem CID | 11737525 |
IUPAC Name | tert-butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCN(CC1)C1=CN=C(N)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15440638
|
Thermo Scientific Alfa Aesar
H54315.MD |
250mg |
1005.00 SEK
250mg |
Estimated Shipment: 22-06-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15420638
|
Thermo Scientific Alfa Aesar
H54315.03 |
1g |
2942.41 SEK
1g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15430638
|
Thermo Scientific Alfa Aesar
H54315.06 |
5g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4-(6-Amino-3-pyridyl)-1-Boc-piperazine is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
571188-59-5 | |
278.36 | |
RMULRXHUNOVPEI-UHFFFAOYSA-N | |
11737525 | |
CC(C)(C)OC(=O)N1CCN(CC1)C1=CN=C(N)C=C1 |
C14H22N4O2 | |
MFCD11594962 | |
tert-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, 1-boc-4-6-aminopyridin-3-yl piperazine, tert-butyl 4-6-amino-3-pyridyl piperazine-1-carboxylate, 1-boc-4-6-amino-3-pyridyl piperazine, 4-6-amino-3-pyridyl-1-boc-piperazine, 1-piperazinecarboxylic acid, 4-6-amino-3-pyridinyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-6-amino-3-pyridyl piperazine, t-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, tert-butyl4-6-aminopyridin-3-yl piperazine-1-carboxylate, 4-6-amino-3-pyridyl piperazine-1-carboxylic acid tert-butyl ester | |
tert-butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate |
Specifications
4-(6-Amino-3-pyridyl)-1-Boc-piperazine | |
C14H22N4O2 | |
10173494 | |
250mg | |
RMULRXHUNOVPEI-UHFFFAOYSA-N | |
tert-butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate | |
11737525 | |
97% |
571188-59-5 | |
MFCD11594962 | |
tert-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, 1-boc-4-6-aminopyridin-3-yl piperazine, tert-butyl 4-6-amino-3-pyridyl piperazine-1-carboxylate, 1-boc-4-6-amino-3-pyridyl piperazine, 4-6-amino-3-pyridyl-1-boc-piperazine, 1-piperazinecarboxylic acid, 4-6-amino-3-pyridinyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-6-amino-3-pyridyl piperazine, t-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, tert-butyl4-6-aminopyridin-3-yl piperazine-1-carboxylate, 4-6-amino-3-pyridyl piperazine-1-carboxylic acid tert-butyl ester | |
Air sensitive | |
CC(C)(C)OC(=O)N1CCN(CC1)C1=CN=C(N)C=C1 | |
278.36 | |
278.35 |
RUO – Research Use Only