Läs mer
4,6-Di-tert-butyl-m-cresol, TRC
Produktkod. 30133915
Change view
Kvantitet:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133915 50 g
17983815 25 g
17911015 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133915 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD42821850G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 497-39-2
Molekylformel C15H24O
Molekylvikt (g/mol) 220.35
Synonym 2,4-Bis(1,1-dimethylethyl)-5-methylphenol, 2,4-Di-tert-butyl-5-methylphenol, 3-Methyl-4,6-di-tert-butylphenol, 4,6-Di-tert-butyl-3-methylphenol, DBMC, Di-tert-Butyl-m-cresol
LEDER Cc1cc(O)c(cc1C(C)(C)C)C(C)(C)C
Specifikationer
Kemiskt namn eller material 4,6-Di-tert-butyl-m-cresol
Kvantitet 50 g
InChI formel InChI=1S/C15H24O/c1-10-8-13(16)12(15(5,6)7)9-11(10)14(2,3)4/h8-9,16H,1-7H3
Formel vikt 220.1827