Läs mer
4,6-Dichloro-N-(imidazolidin-2-ylidene)-2-methylpyrimidin-5-amine (6-Chloromoxonidine), Mikromol™
Produktkod. 30052005
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052005 25 mg
30051040 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052005 Leverantör Mikromol Leverantörsnummer MM1278.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4,6-Dichloro-N-(imidazolidin-2-ylidene)-2-methylpyrimidin-5-amine (6-Chloromoxonidine)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Chloromoxonidine
CAS 352457-35-3
Analyt- eller komponentnamn 6-Chloromoxonidine
Frakt skick Room Temperature
Molekylformel C8 H9 Cl2 N5
Certifikat och efterlevnad ISO 9001
Synonym 4,6-Dichloro-N-(4,5-dihydro-1H-imidazol-2-yl)-2-methyl-5-pyrimidinamine, 6-Chloromoxonidine, Moxonidine Imp. A (EP), Moxonidine Impurity A
InChI formel InChI=1S/C8H9Cl2N5/c1-4-13-6(9)5(7(10)14-4)15-8-11-2-3-12-8/h2-3H2,1H3,(H2,11,12,15)
LEDER Cc1nc(Cl)c(N=C2NCCN2)c(Cl)n1
IUPAC-namn N-(4,6-dichloro-2-methylpyrimidin-5-yl)imidazolidin-2-imine
Molekylvikt (g/mol) 246.1
Kvantitet 25 mg
Formel vikt 245.0235 g/mol