Läs mer
4,6-Dimethoxy-Beta-methyl-3-phenoxy Ambrisentan (Mixture of Diastereomers), TRC
Produktkod. 30370063
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30370063 5 mg
1 options
Produktkod. 30370063 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4605105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4,6-Dimethoxy-beta-methyl-3-phenoxy Ambrisentan (Mixture of Diastereomers)
CAS 159308-03-9
Molekylformel C22H22N2O6
Kvantitet 5 mg
Synonym α-[(4,6-Dimethoxy-2-pyrimidinyl)oxy]-β-methyl-β-phenoxybenzenepropanoic Acid
InChI formel InChI=1S/C22H22N2O6/c1-22(15-10-6-4-7-11-15,30-16-12-8-5-9-13-16)19(20(25)26)29-21-23-17(27-2)14-18(24-21)28-3/h4-14,19H,1-3H3,(H,25,26)
LEDER COC1=CC(OC)=NC(OC(C(O)=O)C(C2=CC=CC=C2)(C)OC3=CC=CC=C3)=N1
Molekylvikt (g/mol) 410.42