Läs mer
4,6-Dimethyl-8-tert-butylcoumarin, TRC
Produktkod. 30374070
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30374070 10 g
30376538 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30374070 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD47143510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17874-34-9
Molekylformel C15 H18 O2
Molekylvikt (g/mol) 230.3
Synonym Coumarin, 8-tert-butyl-4,6-dimethyl- (8CI), 8-(1,1-Dimethylethyl)-4,6-dimethyl-2H-1-benzopyran-2-one, 4,6-Dimethyl-8-tert-butylcoumarin, 8-(tert-Butyl)-4,6-dimethyl-2H-chromen-2-one
IUPAC-namn 8-tert-butyl-4,6-dimethylchromen-2-one
LEDER CC1=CC(=O)Oc2c1cc(C)cc2C(C)(C)C
Specifikationer
Kemiskt namn eller material 4,6-Dimethyl-8-tert-butylcoumarin
Kvantitet 10 g
InChI formel InChI=1S/C15H18O2/c1-9-6-11-10(2)8-13(16)17-14(11)12(7-9)15(3,4)5/h6-8H,1-5H3
Formel vikt 230.1307