Läs mer
4,7-seco-Tebipenemoic Acid Methyl Ester Pivoxil, TRC
Produktkod. 30414522
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30414522 25 mg
30419048 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30414522 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT01286025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391053-29-4
Molekylformel C23 H35 N3 O7 S2
Molekylvikt (g/mol) 529.67
IUPAC-namn 2,2-dimethylpropanoyloxymethyl (2S,3R)-4-[1-(4,5-dihydro-1,3-thiazol-2-yl)azetidin-3-yl]sulfanyl-2-[(2S,3R)-3-hydroxy-1-methoxy-1-oxobutan-2-yl]-3-methyl-2,3-dihydro-1H-pyrrole-5-carboxylate
LEDER COC(=O)[C@H]([C@@H](C)O)[C@@H]1NC(=C(SC2CN(C2)C3=NCCS3)[C@@H]1C)C(=O)OCOC(=O)C(C)(C)C
Specifikationer
Kemiskt namn eller material 4,7-seco-Tebipenemoic Acid Methyl Ester Pivoxil
Kvantitet 25 mg
InChI formel InChI=1S/C23H35N3O7S2/c1-12-16(15(13(2)27)19(28)31-6)25-17(20(29)32-11-33-21(30)23(3,4)5)18(12)35-14-9-26(10-14)22-24-7-8-34-22/h12-16,25,27H,7-11H2,1-6H3/t12-,13-,15-,16-/m1/s1
Formel vikt 529.192