missing translation for 'onlineSavingsMsg'
Learn More

4-(8-Quinolinylsulfonylamino)benzoic acid, 96%, Thermo Scientific™

Catalog Number p-7032363
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
116834-64-1
C16H12N2O4S
328.34
MFCD00601229
NBQVTLMGAPBCJA-UHFFFAOYSA-N
4-quinoline-8-sulfonamido benzoic acid, 4-8-quinolinylsulfonyl amino benzoic acid, benzoicacid, 4-8-quinolinylsulfonyl amino, 4-8-quinolinylsulfonylamino benzoic acid, acmc-20mmuc, cbmicro_030879, cambridge id 5760668, 4-8-quinolylsulfonyl amino benzoic acid, 4-quinolin-8-ylsulfonylamino benzoic acid, 4-quinoline-8-sulfonylamino-benzoic acid
776645
4-(quinolin-8-ylsulfonylamino)benzoic acid
OC(=O)C1=CC=C(NS(=O)(=O)C2=C3N=CC=CC3=CC=C2)C=C1
This item is not returnable. View return policy
116834-64-1
C16H12N2O4S
328.34
MFCD00601229
NBQVTLMGAPBCJA-UHFFFAOYSA-N
4-quinoline-8-sulfonamido benzoic acid, 4-8-quinolinylsulfonyl amino benzoic acid, benzoicacid, 4-8-quinolinylsulfonyl amino, 4-8-quinolinylsulfonylamino benzoic acid, acmc-20mmuc, cbmicro_030879, cambridge id 5760668, 4-8-quinolylsulfonyl amino benzoic acid, 4-quinolin-8-ylsulfonylamino benzoic acid, 4-quinoline-8-sulfonylamino-benzoic acid
776645
4-(quinolin-8-ylsulfonylamino)benzoic acid
OC(=O)C1=CC=C(NS(=O)(=O)C2=C3N=CC=CC3=CC=C2)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 260°C to 269°C
Quantity 250 mg
Formula Weight 328.34
Percent Purity 96%
Chemical Name or Material 4-(8-Quinolinylsulfonylamino)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'