Läs mer
4-Acetamidophenyl Beta-D-Glucuronic Acid Methyl Ester, TRC
Produktkod. 30322854
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30322854 250 mg
1 options
Produktkod. 30322854 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA158495250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Acetamidophenyl beta-D-Glucuronic Acid, Methyl Ester
CAS 570394-17-1
Molekylformel C15 H19 N O8
Kvantitet 250 mg
Synonym 4-Acetamidophenyl -B-D-Glucuronic Acid Methyl Ester
InChI formel InChI=1S/C15H19NO8/c1-7(17)16-8-3-5-9(6-4-8)23-15-12(20)10(18)11(19)13(24-15)14(21)22-2/h3-6,10-13,15,18-20H,1-2H3,(H,16,17)/t10-,11-,12+,13-,15+/m0/s1
LEDER COC(=O)[C@H]1O[C@@H](Oc2ccc(NC(=O)C)cc2)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC-namn methyl (2S,3S,4S,5R,6S)-6-(4-acetamidophenoxy)-3,4,5-trihydroxyoxane-2-carboxylate
Molekylvikt (g/mol) 341.31
Formel vikt 341.1111