Läs mer
4-Acetyl-L-phenylalanine Hydrochloride, TRC
Produktkod. 30324526
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30324526 1 g
1 options
Produktkod. 30324526 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1875401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Acetyl-L-phenylalanine Hydrochloride
CAS 20299-31-4
Molekylformel C11H13NO3 • HCl
Kvantitet 1 g
Synonym p-Acetyl-L-phenylalanine Hydrochloride, L-4-Acetylphenylalanine Hydrochloride
InChI formel InChI=1S/C11H13NO3.ClH/c1-7(13)9-4-2-8(3-5-9)6-10(12)11(14)15;/h2-5,10H,6,12H2,1H3,(H,14,15);1H/t10-;/m0./s1
LEDER CC(=O)C1=CC=C(C=C1)C[C@@H](C(=O)O)N.Cl
IUPAC-namn (2S)-3-(4-acetylphenyl)-2-aminopropanoic acid;hydrochloride
Molekylvikt (g/mol) 207.233646
Formel vikt 243.0662