Learn More
Alfa Aesar™ 4-[Acetyl(methyl)amino]-1-(Boc-amino)-4-phenylpiperidine, 97%
Brand:  Alfa Aesar™ H63967.03
UNSPSC : 12352115
Additional Details : CAS Number : 182621-53-0 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15478606
  • 10620.30 SEK / 1g
Estimated Shipment date
from Supplier 04-10-2017
Add to basket
Specifications
Chemical Identifiers
4-[Acetyl(methyl)amino]-1-(Boc-amino)-4-phenylpiperidine
332.45
tert-Butyl 4-[acetyl(methyl)amino]-4-phenylpiperidine-1-carbamate
182621-53-0
MFCD04971997
Specifications
4-[Acetyl(methyl)amino]-1-(Boc-amino)-4-phenylpiperidine
Solid
332.45
tert-Butyl 4-[acetyl(methyl)amino]-4-phenylpiperidine-1-carbamate
182621-53-0
97%
MFCD04971997
1g
Documentation
Technical Documents