Learn More
4-Acetylphenyl isothiocyanate, 98%, Thermo Scientific™
Catalog Number p-7045771
Click to view available options
Quantity:
1 g
5 g
2131-57-9
C9H7NOS
177.221
MFCD00041100
KAKXOESNHIUYJT-UHFFFAOYSA-N
4-acetylphenyl isothiocyanate, 1-4-isothiocyanatophenyl ethanone, 4-acetylphenylisothiocyanate, 1-4-isothiocyanatophenyl ethan-1-one, acmc-20ampw, bromobenzyl-4-isothiocyanate, 4'-isothiocyanatoacetophenone, 4-acetylphenyl-isothiocyanate, phenylisothiocyanate, 4-acetyl, 1-4-isothiocyanato-phenyl-ethanone
242779
1-(4-isothiocyanatophenyl)ethanone
CC(=O)C1=CC=C(C=C1)N=C=S
2131-57-9
C9H7NOS
177.221
MFCD00041100
KAKXOESNHIUYJT-UHFFFAOYSA-N
4-acetylphenyl isothiocyanate, 1-4-isothiocyanatophenyl ethanone, 4-acetylphenylisothiocyanate, 1-4-isothiocyanatophenyl ethan-1-one, acmc-20ampw, bromobenzyl-4-isothiocyanate, 4'-isothiocyanatoacetophenone, 4-acetylphenyl-isothiocyanate, phenylisothiocyanate, 4-acetyl, 1-4-isothiocyanato-phenyl-ethanone
242779
1-(4-isothiocyanatophenyl)ethanone
CC(=O)C1=CC=C(C=C1)N=C=S
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Melting Point 77°C to 81°C
Boiling Point 112°C (6 mmHg)
Odor Irritating
Quantity 1 g
UN Number UN2923
Beilstein 2207978
Sensitivity Moisture sensitive
Formula Weight 177.23
Percent Purity 98%
Chemical Name or Material 4-Acetylphenyl isothiocyanate