Läs mer
4-Allyl-2-methoxyphenyl Methanesulfonate, TRC
Produktkod. 30324394
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30324394 5 mg
1 options
Produktkod. 30324394 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5525355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Allyl-2-methoxyphenyl Methanesulfonate
CAS 91971-63-0
Molekylformel C11H14O4S
Kvantitet 5 mg
Synonym 4-Allyl-2-methoxyphenol Methanesulfonate, 2-Methoxy-4-(2-propen-1-yl)phenol 1-Methanesulfonate, 2-Methoxy-4-(2-propenyl)phenol Methanesulfonate
InChI formel InChI=1S/C11H14O4S/c1-4-5-9-6-7-10(11(8-9)14-2)15-16(3,12)13/h4,6-8H,1,5H2,2-3H3
LEDER COC1=C(C=CC(CC=C)=C1)OS(C)(=O)=O
IUPAC-namn 4-allyl-2-methoxyphenyl methanesulfonate
Molekylvikt (g/mol) 242.29
Formel vikt 242.06