Learn More
4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 2140-61-6 |
| Molecular Formula | C10H15N3O5 |
| MDL Number | MFCD00006548 |
| Quantity | 10 g |
| SMILES | O=C1N=C(N)C(C)=CN1[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O |
| Molecular Weight (g/mol) | 257.24 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one |