Learn More
4-Amino-1-((2S,3R,4R,5S)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 69-74-9 |
| Molecular Formula | C9H14ClN3O5 |
| MDL Number | MFCD00012839 |
| Quantity | 1 g |
| SMILES | O=C1N=C(N)C=CN1[C@@H]2[C@@H]([C@H]([C@H](CO)O2)O)O.[H]Cl |
| Molecular Weight (g/mol) | 279.68 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Amino-1-((2S,3R,4R,5S)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride |