Läs mer
4-Amino-1-propylpyrrolidin-2-one Hydrochloride, TRC
Produktkod. 30331482
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30331482 500 mg
1 options
Produktkod. 30331482 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA630930500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Amino-1-propylpyrrolidin-2-one Hydrochloride
CAS 1255717-37-3
Molekylformel C7H14N2O . HCl
Kvantitet 500 mg
Synonym 2-Pyrrolidinone, 4-amino-1-propyl-, hydrochloride (1:1) (ACI), 4-Amino-1-propylpyrrolidin-2-one hydrochloride, 4-Amino-1-propyl-2-pyrrolidinone hydrochloride
InChI formel InChI=1S/C7H14N2O.ClH/c1-2-3-9-5-6(8)4-7(9)10;/h6H,2-5,8H2,1H3;1H
LEDER Cl.CCCN1CC(N)CC1=O
IUPAC-namn 4-amino-1-propylpyrrolidin-2-one;hydrochloride
Molekylvikt (g/mol) 178.66
Formel vikt 178.0873