missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-tert-butyl-3-(1’-naphthyl)pyrazolo[3,4-d]pyrimidine, TRC

Product Code. 30317925
Change view
Click to view available options
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30317925 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317925 Supplier Toronto Research Chemicals Supplier No. TRCA6030045MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 4-Amino-1-tert-butyl-3-(1'-naphthyl)pyrazolo[3,4-d]pyrimidine
CAS 221243-82-9
Anteckningar om renhetsgrad HPLC
Molekylformel C19 H19 N5
Kvantitet 5 mg
Synonym 1-(1,1-Dimethylethyl)-3-(1-naphthalenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine, 1-NA-PP1
InChI formel InChI=1S/C19H19N5/c1-19(2,3)24-18-15(17(20)21-11-22-18)16(23-24)14-10-6-8-12-7-4-5-9-13(12)14/h4-11H,1-3H3,(H2,20,21,22)
LEDER CC(C)(C)n1nc(c2cccc3ccccc23)c4c(N)ncnc14
IUPAC-namn 1-tert-butyl-3-naphthalen-1-ylpyrazolo[3,4-d]pyrimidin-4-amine
Molekylvikt (g/mol) 317.39
Formel vikt 317.164
Procent renhet >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.