Läs mer
4-Amino-2-methyl-1-naphthalenol Hydrochloride, TRC
Produktkod. 30326104
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30326104 500 mg
1 options
Produktkod. 30326104 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA616905500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Amino-2-methyl-1-naphthalenol Hydrochloride
CAS 130-24-5
Molekylformel C11 H11 N O . Cl H
Kvantitet 500 mg
Synonym 4-Amino-2-methyl-1-naphthol Hydrochloride, 2-Methyl-4-amino-1-naphthol Hydrochloride, 4-Amino-2-methylnaphthol Hydrochloride, Kayvisyn, Synkamin Hydrochloride, Vitamin K5 Hydrochloride
InChI formel InChI=1S/C11H11NO.ClH/c1-7-6-10(12)8-4-2-3-5-9(8)11(7)13;/h2-6,13H,12H2,1H3;1H
LEDER Cl.Cc1cc(N)c2ccccc2c1O
IUPAC-namn 4-amino-2-methylnaphthalen-1-ol;hydrochloride
Molekylvikt (g/mol) 209.67
Formel vikt 209.0607