Läs mer
(4-Amino-2-methylpyrimidin-5-yl)methyl Dihydrogen Phosphate, TRC
Produktkod. 30317865
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30317865 50 mg
1 options
Produktkod. 30317865 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA61817450MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4-Amino-2-methylpyrimidin-5-yl)methyl Dihydrogen Phosphate
CAS 945-94-8
Molekylformel C6 H10 N3 O4 P
Kvantitet 50 mg
InChI formel InChI=1S/C6H10N3O4P/c1-4-8-2-5(6(7)9-4)3-13-14(10,11)12/h2H,3H2,1H3,(H2,7,8,9)(H2,10,11,12)
LEDER Cc1ncc(COP(=O)(O)O)c(N)n1
IUPAC-namn (4-amino-2-methylpyrimidin-5-yl)methyl dihydrogen phosphate
Molekylvikt (g/mol) 219.1351
Formel vikt 219.0409