Läs mer
4-Amino-3-(4-fluorophenyl)butanoic Acid Hydrochloride, TRC
Produktkod. 30322740
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30322740 5 mg
1 options
Produktkod. 30322740 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6086505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Amino-3-(4-fluorophenyl)butanoic Acid Hydrochloride
CAS 1858241-03-8
Molekylformel C10 H12 F N O2 . H Cl
Kvantitet 5 mg
Synonym β-(Aminomethyl)-4-fluorobenzenepropanoic Acid Hydrochloride, (±)-β-p-Fluorophenyl-GABA Hydrochloride, 4-Amino-3-(4-fluorophenyl)butanoic Acid Hydrochloride
InChI formel InChI=1S/C10H12FNO2.ClH/c11-9-3-1-7(2-4-9)8(6-12)5-10(13)14;/h1-4,8H,5-6,12H2,(H,13,14);1H
LEDER Cl.NCC(CC(=O)O)c1ccc(F)cc1
IUPAC-namn 4-amino-3-(4-fluorophenyl)butanoic acid;hydrochloride
Molekylvikt (g/mol) 233.67
Formel vikt 233.0619