missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-fluorophenylboronic acid hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-4602011
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1256355-32-4
C6H7BFNO2·ClH
191.4
NXCNVCKJELIVDM-UHFFFAOYSA-N
4-amino-3-fluorophenylboronic acid hydrochloride, 4-amino-3-fluorophenylboronic acid, hydrochloride, 4-amino-3-fluorophenyl boronic acid hydrochloride, 4-amino-3-fluorophenylboronic acid,hydrochloride, 4-amino-3-fluorophenylboronic acid,hcl, 4-amino-3-fluorophenylboronic acid, hcl, 4-amino-3-fluorophenyl boronic acid-hydrogen chloride 1/1, boronic acid, b-4-amino-3-fluorophenyl-, hydrochloride 1:1
46738020
This item is not returnable. View return policy
1256355-32-4
C6H7BFNO2·ClH
191.4
NXCNVCKJELIVDM-UHFFFAOYSA-N
4-amino-3-fluorophenylboronic acid hydrochloride, 4-amino-3-fluorophenylboronic acid, hydrochloride, 4-amino-3-fluorophenyl boronic acid hydrochloride, 4-amino-3-fluorophenylboronic acid,hydrochloride, 4-amino-3-fluorophenylboronic acid,hcl, 4-amino-3-fluorophenylboronic acid, hcl, 4-amino-3-fluorophenyl boronic acid-hydrogen chloride 1/1, boronic acid, b-4-amino-3-fluorophenyl-, hydrochloride 1:1
46738020

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 191.4
Percent Purity 97%
Chemical Name or Material 4-Amino-3-fluorophenylboronic acid hydrochloride
compliance-icons
Product Identifier
  • 4-Amino-3-fluorophenylboronic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'