missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitrophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4602016
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
89466-07-9
C6H7BN2O4
181.94
MFCD07437851
IRTXQNNJQZNKRP-UHFFFAOYSA-N
4-amino-3-nitrophenylboronic acid, 4-amino-3-nitrophenylboronicacid, 4-amino-3-nitrophenyl boronic acid, boronic acid,b-4-amino-3-nitrophenyl, pubchem4026, acmc-209r1c, irtxqnnjqznkrp-uhfffaoysa-n, 4-amino-3-nitro-phenyl boronic acid, 4-azanyl-3-nitro-phenyl boronic acid, boronic acid, b-4-amino-3-nitrophenyl
16217915
(4-amino-3-nitrophenyl)boronic acid
NC1=CC=C(C=C1[N+]([O-])=O)B(O)O
This item is not returnable. View return policy
89466-07-9
C6H7BN2O4
181.94
MFCD07437851
IRTXQNNJQZNKRP-UHFFFAOYSA-N
4-amino-3-nitrophenylboronic acid, 4-amino-3-nitrophenylboronicacid, 4-amino-3-nitrophenyl boronic acid, boronic acid,b-4-amino-3-nitrophenyl, pubchem4026, acmc-209r1c, irtxqnnjqznkrp-uhfffaoysa-n, 4-amino-3-nitro-phenyl boronic acid, 4-azanyl-3-nitro-phenyl boronic acid, boronic acid, b-4-amino-3-nitrophenyl
16217915
(4-amino-3-nitrophenyl)boronic acid
NC1=CC=C(C=C1[N+]([O-])=O)B(O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 181.94
Percent Purity 97%
Chemical Name or Material 4-Amino-3-nitrophenylboronic acid
compliance-icons
Product Identifier
  • 4-Amino-3-nitrophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'