missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5,6-dichloropyrimidine, 98%, Thermo Scientific™

Catalog Number p-7055965
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
310400-38-5
C4H3Cl2N3
163.99
MFCD09837293
ISTJUVRUUBNZNB-UHFFFAOYSA-N
4-amino-5,6-dichloropyrimidine, 5,6-dichloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro, 5,6-dichloro-4-pyrimidinamine, 5,6-dichloropyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro-9ci
24219262
5,6-dichloropyrimidin-4-amine
NC1=C(Cl)C(Cl)=NC=N1
This item is not returnable. View return policy
310400-38-5
C4H3Cl2N3
163.99
MFCD09837293
ISTJUVRUUBNZNB-UHFFFAOYSA-N
4-amino-5,6-dichloropyrimidine, 5,6-dichloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro, 5,6-dichloro-4-pyrimidinamine, 5,6-dichloropyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro-9ci
24219262
5,6-dichloropyrimidin-4-amine
NC1=C(Cl)C(Cl)=NC=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White
Quantity 1 g
Beilstein 8678505
Solubility Information Slightly soluble in water.
Formula Weight 163.99
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Amino-5,6-dichloropyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'