Läs mer
4-Amino-5-chloro-2,3-dihydro-7-benzofurancarboxylic Acid, TRC
Produktkod. 30138534
Change view
Kvantitet:
2.5 g
5 g
25 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30138534 25 g -
30137880 5 g -
30007974 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138534 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA60397025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 123654-26-2
Molekylformel C9H8ClNO3
Molekylvikt (g/mol) 213.62
Synonym 4-Amino-5-chloro-2,3-dihydrobenzo[b]furan-7-carboxylic Acid, Prucalopride Impurity
IUPAC-namn 4-amino-5-chloro-2,3-dihydro-1-benzofuran-7-carboxylic acid
LEDER Nc1c(Cl)cc(C(=O)O)c2OCCc12
Specifikationer
Kemiskt namn eller material 4-Amino-5-chloro-2,3-dihydro-7-benzofurancarboxylic Acid
Kvantitet 25 g
InChI formel InChI=1S/C9H8ClNO3/c10-6-3-5(9(12)13)8-4(7(6)11)1-2-14-8/h3H,1-2,11H2,(H,12,13)
Formel vikt 213.0193