missing translation for 'onlineSavingsMsg'
Läs mer

4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-hydroxybenzamide, Mikromol™

Produktkod. 17848400
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
17848400 25 mg
17357079 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17848400 Supplier Mikromol Supplier No. MM0073.060025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-hydroxybenzamide
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-hydroxybenzamide
CAS 38339-95-6
Analyt- eller komponentnamn 4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-hydroxybenzamide
Frakt skick Room Temperature
Molekylformel C13 H20 Cl N3 O2
Certifikat och efterlevnad ISO 17025
Synonym 4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-hydroxybenzamide, Metoclopramide Imp. F (EP), Metoclopramide Hydrochloride Monohydrate Impurity F, Metoclopramide Impurity F, Metoclopramide Impurity F
InChI formel InChI=1S/C13H20ClN3O2/c1-3-17(4-2)6-5-16-13(19)9-7-10(14)11(15)8-12(9)18/h7-8,18H,3-6,15H2,1-2H3,(H,16,19)
LEDER CCN(CC)CCNC(=O)c1cc(Cl)c(N)cc1O
IUPAC-namn 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-hydroxybenzamide
Molekylvikt (g/mol) 285.77
Kvantitet 25 mg
Formel vikt 285.1244 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.