Läs mer
4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic Acid, Mikromol™
Produktkod. 17997314
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17997314 100 mg
17948964 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17997314 Leverantör Mikromol Leverantörsnummer MM0793.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic Acid
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic Acid
CAS 71675-87-1
Analyt- eller komponentnamn 4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic Acid
Frakt skick Room Temperature
Molekylformel C10 H13 N O5 S
Certifikat och efterlevnad ISO 17025
Synonym 4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic Acid, Amisulpride Imp. E (EP), Amisulpride Impurity E
InChI formel InChI=1S/C10H13NO5S/c1-3-17(14,15)9-4-6(10(12)13)8(16-2)5-7(9)11/h4-5H,3,11H2,1-2H3,(H,12,13)
LEDER CCS(=O)(=O)c1cc(C(=O)O)c(OC)cc1N
IUPAC-namn 4-amino-5-ethylsulfonyl-2-methoxybenzoic acid
Molekylvikt (g/mol) 259.28
Kvantitet 100 mg
Formel vikt 259.0514 g/mol