Läs mer
4-Amino-5-fluoro-1-(2,3-di-O-acetyl-Alpha-D-ribofuranosyl)-2(1H)-pyrimidinone, TRC
Produktkod. 17974215
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17974215 10 mg
17924525 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17974215 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA60953510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2230479-30-6
Molekylformel C13 H16 F N3 O6
Molekylvikt (g/mol) 329.28
Synonym Capecitabine Impurity
IUPAC-namn [(2R,3R,4R,5S)-4-acetoxy-5-(4-amino-5-fluoro-2-oxo-pyrimidin-1-yl)-2-methyl-tetrahydrofuran-3-yl] acetate
LEDER C[C@H]1O[C@@H]([C@H](OC(=O)C)[C@@H]1OC(=O)C)N2C=C(F)C(=NC2=O)N
Specifikationer
Kemiskt namn eller material 4-Amino-5-fluoro-1-(2,3-di-O-acetyl-Alpha-D-ribofuranosyl)-2(1H)-pyrimidinone
Analyt- eller komponentnamn 4-Amino-5-fluoro-1-(2,3-di-O-acetyl-Alpha-D-ribofuranosyl)-2(1H)-pyrimidinone
Frakt skick Room Temperature
InChI formel InChI=1S/C13H16FN3O6/c1-5-9(22-6(2)18)10(23-7(3)19)12(21-5)17-4-8(14)11(15)16-13(17)20/h4-5,9-10,12H,1-3H3,(H2,15,16,20)/t5-,9-,10-,12+/m1/s1
Lösningstyp Neat
Industry Type Forensics
Kvantitet 10 mg
Formel vikt 329.1023