Läs mer
4-Amino-5-(formamidomethyl)-2-methylpyrimidine, TRC
Produktkod. 30012865
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012865 1 g
17952585 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012865 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6096751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1886-34-6
Molekylformel C7 H10 N4 O
Molekylvikt (g/mol) 166.18
Synonym 2-Methyl-4-amino-5-(formylaminomethyl)pyrimidine, 4-Amino-5-((formylamino)methyl)-2-methylpyrimidine, N-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]formamide;
IUPAC-namn N-[(4-amino-2-methylpyrimidin-5-yl)methyl]formamide
LEDER O=CNCC1=CN=C(N=C1N)C
Specifikationer
Kemiskt namn eller material 4-Amino-5-(formamidomethyl)-2-methylpyrimidine
Analyt- eller komponentnamn 4-Amino-5-(formamidomethyl)-2-methylpyrimidine
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C7H10N4O/c1-5-10-3-6(2-9-4-12)7(8)11-5/h3-4H,2H2,1H3,(H,9,12)(H2,8,10,11)
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 1 g
Formel vikt 166.0855
Procent renhet >95%