Läs mer
4-Amino-cis-L-proline Dihydrochloride, TRC
Produktkod. 30316769
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30316769 1 g
1 options
Produktkod. 30316769 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5938801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Amino-cis-L-proline Dihydrochloride
CAS 16257-84-4
Molekylformel C5H12Cl2N2O2
Kvantitet 1 g
Synonym (4S)-4-Amino-L-proline, 4-Amino-L-cis-proline Dihydrochloride, (2S,4S)-4-Aminopyrrolidine-2-carboxylic Acid Dihydrochloride
InChI formel InChI=1S/C5H10N2O2.2ClH/c6-3-1-4(5(8)9)7-2-3;;/h3-4,7H,1-2,6H2,(H,8,9);2*1H/t3-,4-;;/m0../s1
LEDER CCc1c(N)nccc1
IUPAC-namn (2S,4S)-4-aminopyrrolidine-2-carboxylic acid;dihydrochloride
Molekylvikt (g/mol) 203.07