Läs mer
4-Amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-iodo-2-methoxy-benzamide, TRC
Produktkod. 30006663
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006663 5 mg
30009714 25 mg
30007407 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006663 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6094105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 176849-91-5
Molekylformel C15 H22 I N3 O2
Molekylvikt (g/mol) 403.26
Synonym 4-Amino-N-[[(2RS)-1-ethyl-pyrrolidin-2-yl]methyl]-5-iodo-2-methoxy-benzamide, Amisulpride Imp. C (EP)
IUPAC-namn 4-amino-N-[(1-ethylpyrrolidin-2-yl)methyl]-5-iodo-2-methoxybenzamide
LEDER CCN1CCCC1CNC(=O)c2cc(I)c(N)cc2OC
Specifikationer
Kemiskt namn eller material 4-Amino-N-[((2RS)-1-ethyl-pyrrolidin-2-yl)methyl]-5-iodo-2-methoxy-benzamide
Analyt- eller komponentnamn 4-Amino-N-[((2RS)-1-ethyl-pyrrolidin-2-yl)methyl]-5-iodo-2-methoxy-benzamide
Frakt skick Room Temperature
InChI formel InChI=1S/C15H22IN3O2/c1-3-19-6-4-5-10(19)9-18-15(20)11-7-12(16)13(17)8-14(11)21-2/h7-8,10H,3-6,9,17H2,1-2H3,(H,18,20)
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 5 mg
Formel vikt 403.0757