Läs mer
4-Amino-N-(2-methylpropyl)-N-[[(4S,5R)-2-oxo-4-(phenylmethyl)-5-oxazolidinyl]methyl]-benzenesulfonamide, TRC
Produktkod. 30011619
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30011619 2.5 mg 2.5mg
30007199 5 mg -
30005808 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011619 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6173752.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1418639-27-6
Molekylformel C21 H27 N3 O4 S
Molekylvikt (g/mol) 417.52
Synonym Benzenesulfonamide, 4-amino-N-(2-methylpropyl)-N-[[(4S,5R)-2-oxo-4-(phenylmethyl)-5-oxazolidinyl]methyl]-, 4-Amino-N-(2-methylpropyl)-N-[[(4S,5R)-2-oxo-4-(phenylmethyl)-5-oxazolidinyl]methyl]benzenesulfonamide, 4-Amino-N-(2-methylpropyl)-N-[[(4S,5R)-4-benzyl-2-oxooxazolidin-5-yl]methyl]benzenesulfonamide, 4-Amino-N-(2-methylpropyl)-N-[[(4S,5R)-2-oxo-4-(phenylmethyl)oxazolidin-5-yl]methyl]benzenesulfonamide, 4949W94 (GW 288003X), GW 461013X
IUPAC-namn 4-amino-N-[[(4S,5R)-4-benzyl-2-oxo-1,3-oxazolidin-5-yl]methyl]-N-(2-methylpropyl)benzenesulfonamide
LEDER CC(C)CN(C[C@H]1OC(=O)N[C@H]1Cc2ccccc2)S(=O)(=O)c3ccc(N)cc3
Specifikationer
Kemiskt namn eller material 4-Amino-N-(2-methylpropyl)-N-[((4S,5R)-2-oxo-4-(phenylmethyl)oxazolidin-5-yl)methyl]benzenesulfonamide
Analyt- eller komponentnamn 4-Amino-N-(2-methylpropyl)-N-[((4S,5R)-2-oxo-4-(phenylmethyl)oxazolidin-5-yl)methyl]benzenesulfonamide
Frakt skick Room Temperature
InChI formel InChI=1S/C21H27N3O4S/c1-15(2)13-24(29(26,27)18-10-8-17(22)9-11-18)14-20-19(23-21(25)28-20)12-16-6-4-3-5-7-16/h3-11,15,19-20H,12-14,22H2,1-2H3,(H,23,25)/t19-,20+/m0/s1
Lösningstyp Neat
Industry Type Forensics
Kvantitet 2.5 mg
Formel vikt 417.1722