missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzamidine dihydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7058409
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2498-50-2
C7H11Cl2N3
208.086
MFCD00013001
GHEHNICLPWTXJC-UHFFFAOYSA-N
4-aminobenzamidine dihydrochloride, p-aminobenzamidine dihydrochloride, 4-aminobenzenecarboximidamide dihydrochloride, 4-aminobenzimidamide dihydrochloride, 4-aminobenzamidine 2hcl, 4-amidinoaniline dihydrochloride, benzenecarboximidamide, 4-amino-, dihydrochloride, unii-3c7vnu62ag, benzamidine, p-amino-, dihydrochloride, 3c7vnu62ag
75626
CHEBI:10618
4-aminobenzenecarboximidamide;dihydrochloride
C1=CC(=CC=C1C(=N)N)N.Cl.Cl
This item is not returnable. View return policy
2498-50-2
C7H11Cl2N3
208.086
MFCD00013001
GHEHNICLPWTXJC-UHFFFAOYSA-N
4-aminobenzamidine dihydrochloride, p-aminobenzamidine dihydrochloride, 4-aminobenzenecarboximidamide dihydrochloride, 4-aminobenzimidamide dihydrochloride, 4-aminobenzamidine 2hcl, 4-amidinoaniline dihydrochloride, benzenecarboximidamide, 4-amino-, dihydrochloride, unii-3c7vnu62ag, benzamidine, p-amino-, dihydrochloride, 3c7vnu62ag
75626
CHEBI:10618
4-aminobenzenecarboximidamide;dihydrochloride
C1=CC(=CC=C1C(=N)N)N.Cl.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Quantity 1 g
Beilstein 3692927
Sensitivity Hygroscopic
Formula Weight 208.09
Percent Purity 97%
Chemical Name or Material 4-Aminobenzamidine dihydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'