missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzamidoxime, 97%, Thermo Scientific™

771.78 SEK - 2722.12 SEK

Chemical Identifiers

CAS 277319-62-7
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.169
MDL Number MFCD08061151
InChI Key CNFNMMJKXWOLPY-UHFFFAOYSA-N
Synonym 4-amino-n'-hydroxybenzimidamide, 4-amino-n'-hydroxybenzene-1-carboximidamide, z-4-amino-n'-hydroxybenzenecarboximidamide
PubChem CID 5478941
IUPAC Name 4-amino-N'-hydroxybenzenecarboximidamide
SMILES C1=CC(=CC=C1C(=NO)N)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15428247
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
H51788.03
1g
771.78 SEK
1g
View alternative products
Save Up to 
»
In stock, ready to ship: 1
Please to purchase this item. Need a web account? Register with us today!
15438247
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
H51788.06
5g
2722.12 SEK
5g
View alternative products
Save Up to 
»
Estimated Shipment: 05-01-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

277319-62-7
151.169
CNFNMMJKXWOLPY-UHFFFAOYSA-N
5478941
C1=CC(=CC=C1C(=NO)N)N
C7H9N3O
MFCD08061151
4-amino-n'-hydroxybenzimidamide, 4-amino-n'-hydroxybenzene-1-carboximidamide, z-4-amino-n'-hydroxybenzenecarboximidamide
4-amino-N'-hydroxybenzenecarboximidamide

Specifications

4-Aminobenzamidoxime
168°C to 170°C
MFCD08061151
1g
C1=CC(=CC=C1C(=NO)N)N
151.169
151.17
277319-62-7
C7H9N3O
4-amino-n'-hydroxybenzimidamide, 4-amino-n'-hydroxybenzene-1-carboximidamide, z-4-amino-n'-hydroxybenzenecarboximidamide
CNFNMMJKXWOLPY-UHFFFAOYSA-N
4-amino-N'-hydroxybenzenecarboximidamide
5478941
97%
SDS
Product Certifications

RUO – Research Use Only