missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-7021431
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
1137-41-3
C13H11NO
197.24
MFCD00007895
RBKHNGHPZZZJCI-UHFFFAOYSA-N
4-aminobenzophenone, 4-aminophenyl phenyl methanone, 4-benzoylaniline, p-aminobenzophenone, methanone, 4-aminophenyl phenyl, benzophenone, 4-amino, p-benzoylaniline, usaf a-233, 4-aminophenyl-phenylmethanone, 4-aminophenyl-phenyl-methanone
14346
(4-aminophenyl)-phenylmethanone
NC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
This item is not returnable. View return policy
1137-41-3
C13H11NO
197.24
MFCD00007895
RBKHNGHPZZZJCI-UHFFFAOYSA-N
4-aminobenzophenone, 4-aminophenyl phenyl methanone, 4-benzoylaniline, p-aminobenzophenone, methanone, 4-aminophenyl phenyl, benzophenone, 4-amino, p-benzoylaniline, usaf a-233, 4-aminophenyl-phenylmethanone, 4-aminophenyl-phenyl-methanone
14346
(4-aminophenyl)-phenylmethanone
NC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

4-Aminobenzophenone is associated with 4-benzoylpyridine and it is used as a frequency doubler for semiconductor lasers. It is used in the preparation of 1-[4-[N,N-bis(trimethylsilyl)amino]phenyl]-1-phenylethylene. It is used as a precursor to prepare 1,2,3-triazoles, pyrimidotriazoles, benzothiazole and pyrimidinone derivatives. It is involved in the formation of coordination complexes with chromium(III).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzophenone is associated with 4-benzoylpyridine and it is used as a frequency doubler for semiconductor lasers. It is used in the preparation of 1-[4-[N,N-bis(trimethylsilyl)amino]phenyl]-1-phenylethylene. It is used as a precursor to prepare 1,2,3-triazoles, pyrimidotriazoles, benzothiazole and pyrimidinone derivatives. It is involved in the formation of coordination complexes with chromium(III).

Solubility
Soluble in cold water, alcohol and ether.

Notes
Air and light sensitive. Incompatible with strong acids.
TRUSTED_SUSTAINABILITY
Melting Point 121°C to 124°C
Boiling Point 245°C to 247°C (22 mmHg)
Quantity 10 g
Beilstein 389292
Solubility Information Soluble in cold water,alcohol and ether.
Formula Weight 197.24
Percent Purity 98%
Chemical Name or Material 4-Aminobenzophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'