missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-55926
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
1137-41-3
C13H11NO
197.24
MFCD00007895
RBKHNGHPZZZJCI-UHFFFAOYSA-N
4-aminobenzophenone, 4-aminophenyl phenyl methanone, 4-benzoylaniline, p-aminobenzophenone, methanone, 4-aminophenyl phenyl, benzophenone, 4-amino, p-benzoylaniline, usaf a-233, 4-aminophenyl-phenylmethanone, 4-aminophenyl-phenyl-methanone
14346
(4-aminophenyl)-phenylmethanone
NC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
This item is not returnable. View return policy
1137-41-3
C13H11NO
197.24
MFCD00007895
RBKHNGHPZZZJCI-UHFFFAOYSA-N
4-aminobenzophenone, 4-aminophenyl phenyl methanone, 4-benzoylaniline, p-aminobenzophenone, methanone, 4-aminophenyl phenyl, benzophenone, 4-amino, p-benzoylaniline, usaf a-233, 4-aminophenyl-phenylmethanone, 4-aminophenyl-phenyl-methanone
14346
(4-aminophenyl)-phenylmethanone
NC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 120°C to 124°C
Color Yellow
Boiling Point 246°C (17.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4COC6H5
Quantity 10 g
Beilstein 14, 81
Solubility Information Solubility in water: hardly soluble in cold water. Other solubilities: freely soluble in alcohol and ether
Formula Weight 197.24
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Aminobenzophenone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Aminobenzophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'