Learn More
4-Aminobiphenyl-d9, TRC
Product Code. 30315850
Change view
Click to view available options
Kvantitet:
10 mg
Product Code. Kvantitet
30315850 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315850 Supplier Toronto Research Chemicals Supplier No. TRCA60179710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 4-Aminobiphenyl D9
CAS 344298-96-0
Anteckningar om renhetsgrad HPLC
Molekylformel C12 2H9 H2 N
Kvantitet 10 mg
Synonym [1,1'-Biphenyl-2,2',3,3',4',5,5',6,6'-d9]-4-amine (9CI), 4-Aminobiphenyl D9
InChI formel InChI=1S/C12H11N/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9H,13H2/i1D,2D,3D,4D,5D,6D,7D,8D,9D
LEDER [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2c([2H])c([2H])c(N)c([2H])c2[2H]
IUPAC-namn 2,3,5,6-tetradeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)aniline
Molekylvikt (g/mol) 178.28
Formel vikt 178.1456