Läs mer
4-(Aminomethyl)-1-cyclohexene-1-carboxylic Acid (Tranexamic acid Impurity), TRC
Produktkod. 30323695
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30323695 5 mg
1 options
Produktkod. 30323695 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6155305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (RS)-4-(aminomethyl)cyclohex-1-enecarboxylic acid
CAS 330838-52-3
Molekylformel C8 H13 N O2
Kvantitet 5 mg
Synonym 4-(aminomethyl)-1-Cyclohexene-1-carboxylic acid, USP Tranexamic Acid Related Compound C, (RS)-4-(aminomethyl)cyclohex-1-enecarboxylic acid
InChI formel InChI=1S/C8H13NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h3,6H,1-2,4-5,9H2,(H,10,11)
LEDER NCC1CCC(=CC1)C(=O)O
IUPAC-namn 4-(aminomethyl)cyclohexene-1-carboxylic acid
Molekylvikt (g/mol) 155.19
Formel vikt 155.0946