Läs mer
4-(Aminomethyl)-6,7-dimethoxycoumarin, TRC
Produktkod. 30316067
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30316067 10 mg
30327651 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30316067 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA61587510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(Aminomethyl)-6,7-dimethoxycoumarin
CAS 849042-47-3
Molekylformel C12 H13 N O4
Kvantitet 10 mg
InChI formel InChI=1S/C12H13NO4/c1-15-10-4-8-7(6-13)3-12(14)17-9(8)5-11(10)16-2/h3-5H,6,13H2,1-2H3
LEDER COc1cc2OC(=O)C=C(CN)c2cc1OC
IUPAC-namn 4-(aminomethyl)-6,7-dimethoxychromen-2-one
Molekylvikt (g/mol) 235.24
Formel vikt 235.0845