Läs mer
(4-Aminomethyl)benzyl Alcohol Hydrochloride, TRC
Produktkod. 30319041
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30319041 100 mg
1 options
Produktkod. 30319041 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA613630100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4-Aminomethyl)benzyl Alcohol Hydrochloride
CAS 34403-46-8
Molekylformel C8H11NO . (HCl)
Kvantitet 100 mg
Synonym 4-(Aminomethyl)benzenemethano Hydrochloride, p-(Aminomethyl)benzyl Alcohol Hydrochloride, [4-(Aminomethyl)phenyl]methanol Hydrochloride
InChI formel InChI=1S/C22H26O/c1-4-21(3)12-11-20-19-8-6-15-14-16(23)7-9-17(15)18(19)10-13-22(20,21)5-2/h1,6,8,20H,5,7,9-14H2,2-3H3/t20-,21+,22-/m0/s1
LEDER CC[C@]12CCc3c4CCC(=O)Cc4ccc3[C@@H]1CC[C@@]2(C)C#C
IUPAC-namn (13S,14R,17S)-13-ethyl-17-ethynyl-17-methyl-1,2,4,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
Molekylvikt (g/mol) 173.64
Formel vikt 306.1984