Läs mer
4-(Aminomethyl)cyclohexanecarboxylic Acid, TRC
Produktkod. 30342354
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30342354 5 g
1 options
Produktkod. 30342354 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6349085G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(Aminomethyl)cyclohexanecarboxylic Acid
CAS 701-54-2
Molekylformel C8H15NO2
Kvantitet 5 g
Synonym Cyclohexanecarboxylic acid, 4-(aminomethyl)-, 4-(Aminomethyl)cyclohexanecarboxylic acid, 1-(Aminomethyl)cyclohexane-4-carboxylic acid, 4-(Aminomethyl)-1-cyclohexanecarboxylic acid, AMCA, p-(Aminomethyl)cyclohexanecarboxylic acid
InChI formel InChI=1S/C8H15NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h6-7H,1-5,9H2,(H,10,11)
LEDER NCC1CCC(CC1)C(=O)O
IUPAC-namn 4-(aminomethyl)cyclohexane-1-carboxylic acid
Molekylvikt (g/mol) 157.21
Formel vikt 157.1103